Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S,S,R,S,R)-Boc-Dap-NE | 3026595-14-9 | >98.0% | 420.54 g/mol | C23H36N2O5 | 10 MG
SDP

Catalog No. 5000299110
Encompass_Preferred

(S,S,R,S,R)-Boc-Dap-NE is a Boc-protected diaminopropionic dipeptide isomer used as a drug intermediate in the synthesis of monomethyl auristatin E and related antibody-drug conjugate payloads. Supplied in small research quantities for medicinal chemistry and ADC payload synthesis.

  • Drug intermediate suitable for ADC payload synthesis.
  • Purity greater than 98.0%.
  • Molecular formula C23H36N2O5.
  • Molecular weight 420.54 g/mol.
  • CAS number 3026595-14-9.
  • Soluble in DMSO: 200 mg/mL (ultrasonic); 10 mM in DMSO.
  • Available in small research sizes (10 mg, 25 mg, 100 mg).

Catalog No. 50-002-99110 Supplier Medchemexpress LLC Supplier No. HY78931C10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S,S,R,S,R)-Boc-Dap-NE is a Boc-protected diaminopropionic dipeptide isomer used as a drug intermediate in the synthesis of monomethyl auristatin E and related antibody-drug conjugate payloads. Supplied in small research quantities for medicinal chemistry and ADC payload synthesis.

  • Drug intermediate suitable for ADC payload synthesis.
  • Purity greater than 98.0%.
  • Molecular formula C23H36N2O5.
  • Molecular weight 420.54 g/mol.
  • CAS number 3026595-14-9.
  • Soluble in DMSO: 200 mg/mL (ultrasonic); 10 mM in DMSO.
  • Available in small research sizes (10 mg, 25 mg, 100 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.