Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Relay Solid State 240 V 45A

Catalog No. NC0633822 Shop All Thermo Scientific Products
Encompass_Preferred

Relay Solid State 240 V 45A.

Catalog No. NC0633822 Supplier Thermo Scientific Supplier No. RYX37
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Relay Solid State 240 V 45A.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.