Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S,S,S)-AHPC hydrochloride | 2115897-23-7 | 99.2% | 467.02 | C22H31ClN4O3S | 5MG
SDP

Catalog No. 5000239515
Encompass_Preferred

(S,S,S)-AHPC hydrochloride is the hydrochloride salt of an AHPC-derived VHL ligand used as a building block and negative control in E3 ligase recruitment and PROTAC research. It facilitates the study of VHL-mediated ubiquitination and proteasomal degradation, and is supplied for synthetic and assay applications in small research quantities.

  • High purity 99.2%
  • Molecular weight 467.02
  • Chemical formula C22H31ClN4O3S
  • Solid white to yellow appearance
  • Suitable as a negative control and VHL ligand building block for PROTAC design
  • Available in small milligram pack sizes for research use

Catalog No. 50-002-39515 Supplier Medchemexpress LLC Supplier No. HY125845A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S,S,S)-AHPC hydrochloride is the hydrochloride salt of an AHPC-derived VHL ligand used as a building block and negative control in E3 ligase recruitment and PROTAC research. It facilitates the study of VHL-mediated ubiquitination and proteasomal degradation, and is supplied for synthetic and assay applications in small research quantities.

  • High purity 99.2%
  • Molecular weight 467.02
  • Chemical formula C22H31ClN4O3S
  • Solid white to yellow appearance
  • Suitable as a negative control and VHL ligand building block for PROTAC design
  • Available in small milligram pack sizes for research use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.