Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S,S,S,S,R)-Boc-Dap-NE | 3026594-75-9 | 99.8% | 50 MG
SDP

Catalog No. 5000298416
Encompass_Preferred

(S,S,S,S,R)-Boc-Dap-NE is a Boc-protected diaminopropane derivative used as an intermediate in the synthesis of monomethyl auristatin E (MMAE) and related auristatin analogs. It is supplied as a purified solid for research and medicinal chemistry synthesis.

  • High purity: 99.8%.
  • Molecular formula: C23H36N2O5.
  • Molecular weight: 420.54 g/mol.
  • Physical form: off-white to white powder, stored under inert gas.
  • Common pack sizes: 25 MG, 50 MG, 100 MG (availability may vary).

Catalog No. 50-002-98416 Supplier Medchemexpress LLC Supplier No. HY78931B10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S,S,S,S,R)-Boc-Dap-NE is a Boc-protected diaminopropane derivative used as an intermediate in the synthesis of monomethyl auristatin E (MMAE) and related auristatin analogs. It is supplied as a purified solid for research and medicinal chemistry synthesis.

  • High purity: 99.8%.
  • Molecular formula: C23H36N2O5.
  • Molecular weight: 420.54 g/mol.
  • Physical form: off-white to white powder, stored under inert gas.
  • Common pack sizes: 25 MG, 50 MG, 100 MG (availability may vary).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.