Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S,S,S)-Triglycidyl Isocyanurate 98.0+%, TCI America™
SDP

Catalog No. I05811G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I05811G Supplier TCI America Supplier No. I05811G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 240408-81-5
Molecular Formula C12H15N3O6
Molecular Weight (g/mol) 297.27
MDL Number MFCD03844799
InChI Key OUPZKGBUJRBPGC-UHFFFAOYNA-N
Synonym Isocyanuric Acid (S,S,S)-Tris(2,3-epoxypropyl) Ester, Isocyanuric Acid (S,S,S)-Triglycidyl Ester, (S,S,S)-Tris(2,3-epoxypropyl) Isocyanurate
IUPAC Name tris[(oxiran-2-yl)methyl]-1,3,5-triazinane-2,4,6-trione
SMILES O=C1N(CC2CO2)C(=O)N(CC2CO2)C(=O)N1CC1CO1

Specifications

Melting Point 107°C
Color White
Quantity 1 g
UN Number 2811
Formula Weight 297.27
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material (S,S,S)-Triglycidyl Isocyanurate
EINECSNumber (5)-1052
RTECSNumber XZ1994900
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.