Learn More
Uline Stainless Steel Storage Cabinet - 48 x 24 x 73"
Catalog No. NC2866735
Encompass
Lock up expensive lab equipment and corrosive chemicals. Lasts a lifetime.Welded, heavy-duty construction stands up to continuous use in harsh environments.Type 304 stainless steel will not rust.Shelves adjust in 3 1/2" increments. 500 lb. shelf capacity.Recessed paddle lock handle. Full-height hinges provide smooth closure and longevity.
Catalog No. NC2866735 Supplier Uline Supplier No. H5589
Lock up expensive lab equipment and corrosive chemicals. Lasts a lifetime.Welded, heavy-duty construction stands up to continuous use in harsh environments.Type 304 stainless steel will not rust.Shelves adjust in 3 1/2" increments. 500 lb. shelf capacity.Recessed paddle lock handle. Full-height hinges provide smooth closure and longevity.