Learn More
Next Advance Inc Stainless Steel UFO Beads 5.6 mm, 50 mL
SDP
Catalog No. NC3785830
Encompass
5.6 mm Stainless steel beads in cone ball shape for specialized homogenization- density is 7.9 g/cc, sold in 50 mL package. For the homogenization of tough tissue. Use with 5 mL tubes or 50 mL tubes.
Catalog No. NC3785830 Supplier Next Advance Inc Supplier No. SSUFO56
5.6 mm Stainless steel beads in cone ball shape for specialized homogenization- density is 7.9 g/cc, sold in 50 mL package. For the homogenization of tough tissue. Use with 5 mL tubes or 50 mL tubes.