Learn More
Medchemexpress LLC HY-17518A 10mg Medchemexpress, (S,S)-Valifenalate CAS:283159-94-4 Purity:>98%
SDP
Catalog No. 501969993
Encompass_Preferred
Medchemexpress, HY-17518A 10mg (S,S)-Valifenalate CAS:283159-94-4 (S,S)-Valifenalate ((S,S)-IR5885) is an acylamino acid fungicide and is used to control a wide range of fungi belonging to the class of Oomycetes. (S,S)-Valifenalate ((S,S)-IR5885) interferes with the cell-wall synthesis thus affecting the growth stages of the pathogens controlled, both outside (on the spores) or inside the plant (on the mycelium). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9993 Supplier Medchemexpress LLC Supplier No. HY17518A10MG
Medchemexpress, HY-17518A 10mg (S,S)-Valifenalate CAS:283159-94-4 (S,S)-Valifenalate ((S,S)-IR5885) is an acylamino acid fungicide and is used to control a wide range of fungi belonging to the class of Oomycetes. (S,S)-Valifenalate ((S,S)-IR5885) interferes with the cell-wall synthesis thus affecting the growth stages of the pathogens controlled, both outside (on the spores) or inside the plant (on the mycelium). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.