Learn More
Regis Technologies Inc HPLC Column (S,S) Whelk-O1 3.5µm, 3.0 x 100 mm
SDP
Supplier:  Regis Technologies Inc 1780125300
Encompass
The Whelk-O 1 Chiral Stationary Phase is based on 1-(3,5-Dinitrobenzamido)-1,2,3,4,-tetrahydrophenanthrene. This phase allows separation of nderivatized racemates from a number of families including amides, epoxides, esters, ureas, carbamates, ethers, aziridines, phosphonates, aldehydes, ketones, carboxylic acids, and alcohols. The Whelk-O 1 was originally designed for the separation of underivatized nonsteroidal anti-inflammatory drugs (NSAIDs). This π-electron acceptor/π-electron donor phase exhibits an extraordinary degree of generality, allowing resolution of a wide variety of underivatized racemates. This broad versatility observed on the Whelk-O 1 column, compares favorably with polysaccharide-derived chiral stationary phases. In addition, because of the Whelk-O 1’s covalent nature, this chiral phase is compatible with all commonly used mobile phases, including aqueous systems-a distinct advantage over polysaccharide-derived chiral stationary phases.
Catalog No. NC1619282