Learn More
Regis Technologies Inc (S,S) Whelk-O® 1 Chiral Column, 1.8 µm, 10 cm x 4.6 mm ID
SDP
Supplier:  Regis Technologies Inc 1780161300
Encompass
Chiral HPLC/SFC Column, Pirkle-type Chiral Stationary Phase (CSP). The Whelk-O 1 is useful for the separation of underivatized enantiomers in a number of families including amides, epoxides, esters, ureas, carbamates, ethers, aziridines, phosphonates, aldehydes, ketones, carboxylic acids, alcohols and non-steroidal anti-inflammatory drugs (NSAIDs). This π-electron acceptor/π-electron donor phase exhibits an extraordinary degree of generality. The broad versatility observed on the Whelk-O 1 column compares favorably with polysaccharide-derived chiral stationary phases. In addition, because Whelk-O 1 is covalently bonded to the support, the phase is compatible with all commonly used mobile phases, including aqueous systems — a distinct advantage over coated polysaccharide-derived chiral stationary phases. Other advantages include column durability, excellent efficiency, ability to invert elution order and excellent preparative capacity. Whelk-O 1 is recognized as USP Code L102.
Catalog No. 50-153-2978