Learn More
Uline Deluxe Mobile Stainless Steel Worktable with Bottom Shelf - 48 x 24"
Catalog No. NC2866715
Encompass
Mix, chop and roll. Move your workspace where you need it.Easy portability for reconfiguring and cleaning work area.All stainless steel construction. Type 304 top.1 5/8" diameter steel tube legs. 35" table height.Adjustable bottom shelf. 4" polyurethane locking swivel casters.NSF certified.
Catalog No. NC2866715 Supplier Uline Supplier No. H10292
Mix, chop and roll. Move your workspace where you need it.Easy portability for reconfiguring and cleaning work area.All stainless steel construction. Type 304 top.1 5/8" diameter steel tube legs. 35" table height.Adjustable bottom shelf. 4" polyurethane locking swivel casters.NSF certified.