Learn More
Medchemexpress LLC (S)-S007-1558 | 1327156-14-8 | ≥98.0% | 339.35 g/mol | C17H17N5O3 | 5 MG
SDP
Catalog No. 5000221220
Encompass_Preferred
(S)-S007-1558 is a small-molecule glycoprotein VI (GPVI) platelet receptor antagonist for research use. It inhibits collagen-induced platelet aggregation with a reported IC50 of 0.17 μM in human platelets and is supplied at high purity for biochemical and cellular studies. Recommended cold storage preserves compound stability.
Catalog No. 50-002-21220 Supplier Medchemexpress LLC Supplier No. HY1443905MG
(S)-S007-1558 is a small-molecule glycoprotein VI (GPVI) platelet receptor antagonist for research use. It inhibits collagen-induced platelet aggregation with a reported IC50 of 0.17 μM in human platelets and is supplied at high purity for biochemical and cellular studies. Recommended cold storage preserves compound stability.