Learn More
Medchemexpress LLC HY-129356A 5mg Medchemexpress, (S)-Seco-Duocarmycin SA CAS:152785-82-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY129356A5MG
Encompass_Preferred
Medchemexpress, HY-129356A 5mg (S)-Seco-Duocarmycin SA CAS:152785-82-5 (S)-Seco-Duocarmycin SA is a DNA alkylator, cytotoxic to cancer cells, and acts as a ADC cytotoxin for antibody-drug conjugates. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1202