Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-SEGPHOS(regR) 99.0+%, TCI America™
SDP

Catalog No. S0929200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0929200MG 200 mg
S09291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0929200MG Supplier TCI America Supplier No. S0929200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 210169-54-3
Molecular Formula C38H28O4P2
Molecular Weight (g/mol) 610.586
MDL Number MFCD09753005
InChI Key RZZDRSHFIVOQAF-UHFFFAOYSA-N
Synonym (S)-(-)-5,5′C-Bis(diphenylphosphino)-4,4′C-bi-1,3-benzodioxole, (S)-(-)-4,4′C-Bis(diphenylphosphino)-3,3′C-bi(1,2-methylenedioxybenzene)
PubChem CID 11017510
IUPAC Name [4-(5-diphenylphosphanyl-1,3-benzodioxol-4-yl)-1,3-benzodioxol-5-yl]-diphenylphosphane
SMILES C1OC2=C(O1)C(=C(C=C2)P(C3=CC=CC=C3)C4=CC=CC=C4)C5=C(C=CC6=C5OCO6)P(C7=CC=CC=C7)C8=CC=CC=C8

Specifications

Melting Point 217°C
Quantity 200 mg
Formula Weight 610.59
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-SEGPHOS(regR)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.