Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ S-Series Hood with Inner Shroud, Integrated Head Suspension

Catalog No. 19039549 Shop All 3M Products
Change view
Click to view available options
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-039-549 Small
19-039-548 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-039-549 Supplier 3M™ Supplier No. 7000127463
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Economy hood for use with 3M™ PAPR and SAR systems. Available in small/medium or medium/large size.

  • Versaflo S-Series economy hood
  • Polypropylene
  • Air Regulating Valve, Cooling / Heating Valve, Low Pressure Valve
  • 20/Case
  • Applications include cleaning, assembly, mechanical, painting
  • 3M Catalog# S-403L-20, S-403S-20
TRUSTED_SUSTAINABILITY

Specifications

Assigned Protection Factor (APF) 1000
Certifications/Compliance NIOSH
Color White
Material Polypropylene
Product Type Hood with Inner Shroud
Size Small
For Use With (Application) Military maintenance, repair and operation, pharmaceutical, transportation, metal production and fabrication, automotive, food and beverage manufacturing
For Use With (Equipment) Powered air purifying and supplied air respirator systems
Includes Head, face, neck, and shoulder coverage, fabric suspension, inner shroud
Model S-403S-20
Quantity 20/Cs.
Suspension Type Integrated
Visor Type PETG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.