Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ S-Series Hood with Inner Shroud, Premium Head Suspension

Catalog No. 19805543 Shop All 3M Products
Change view
Click to view available options
Product Type:
Hood with Inner Shroud
Hood with Neck Collar
Includes:
Hood with inner shroud only
S-805 hood with neck collar, S-950 Premium Head Suspension, S-955 Airflow Adjustment Cover, S-954 Size Reducing Pad, S-922 Peel-Off
S-807 Hood with Inner Shroud, S-950 Premium Head Suspension, S-955 Airflow Adjustment Cover, S-954 Size Reducing Pad, S-922 Peel-Off
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Includes
19-805-543 Hood with Inner Shroud S-807 Hood with Inner Shroud, S-950 Premium Head Suspension, S-955 Airflow Adjustment Cover, S-954 Size Reducing Pad, S-922 Peel-Off
17-623-801 Hood with Neck Collar S-805 hood with neck collar, S-950 Premium Head Suspension, S-955 Airflow Adjustment Cover, S-954 Size Reducing Pad, S-922 Peel-Off
19-805-542 Hood with Inner Shroud Hood with inner shroud only
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-805-543 Supplier 3M™ Supplier No. 7000002350
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S-Series hood with sealed seams features a double shroud design for its respiratory seal.

Premium suspension hoods and head covers

  • Head covers provide head and face coverage; hoods cover head, face, and neck
  • Fully assembled and ready to use
  • Suitable for applications that require frequent replacement of the entire headgear assembly, such as pharmaceutical manufacturing, or situations where there is less soiling of the outer fabric
  • Integrated models feature wide face seal with soft elastic for improved comfort and reduced noise

Premium Reusable Suspensions with Hoods

  • Ideal for high disposal rate environments
  • Soiled hood is replaced; suspension and air ducting are reused
  • Rigid ducting for consistent air path
  • Externally adjustable airflow control allows wearer to modify airflow distribution
  • Suspension features independent adjustments for the crown and head circumference; allows custom fit for wide range of head sizes
  • 3M Catalog# S-805-5, S-807-5, S-855, S-857
TRUSTED_SUSTAINABILITY

Specifications

Assigned Protection Factor (APF) 1000
Certifications/Compliance NIOSH
Color Gray
Material Zytron 200
Product Type Hood with Inner Shroud
Size Standard
For Use With (Application) Assembly, facility maintenance, machine operation, painting, sanding
For Use With (Equipment) GVP, Breathe-Easy™, V Series, T-300, T-600, T-800
Includes S-807 Hood with Inner Shroud, S-950 Premium Head Suspension, S-955 Airflow Adjustment Cover, S-954 Size Reducing Pad, S-922 Peel-Off
Model S-857
Quantity 1 Ea.
Suspension Type Premium Resusable Suspension
Visor Type Acetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.