Learn More
Medchemexpress LLC HY-103504 5mg Medchemexpress, (S)-SNAP5114 CAS:157604-55-2 Purity:>98%
SDP
Catalog No. 501932247
Encompass_Preferred
Medchemexpress, HY-103504 5mg (S)-SNAP5114 CAS:157604-55-2 (S)-SNAP5114 is a selective GABA transport inhibitor, with IC 50 values of 5 μM and 21 μM for hGAT-3 and rGAT-2, respectively. (S)-SNAP5114 is an anticonvulsant drug [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2247 Supplier Medchemexpress LLC Supplier No. HY1035045MG
Add to Cart
Add to Cart
Medchemexpress, HY-103504 5mg (S)-SNAP5114 CAS:157604-55-2 (S)-SNAP5114 is a selective GABA transport inhibitor, with IC 50 values of 5 μM and 21 μM for hGAT-3 and rGAT-2, respectively. (S)-SNAP5114 is an anticonvulsant drug [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.