Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(-)-Styrene Oxide

Catalog No. 54010225G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
54-010-225G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 54-010-225G Supplier Sigma Aldrich Supplier No. 54010225G
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Styrene oxide is an oxirane derivative, which is used as a monomer for the synthesis of polyesters with relatively highTg values via ring-opening copolymerization. It can also be used as a useful intermediate in the preparation of levamisole, a nematocideand anticancer agent.(S)-(-)-Styrene oxide can undergo copolymerization with CO2 to form the corresponding polycarbonate in the presence of a cobalt-based catalyst system.Manufactured under license by Sterling Pharma Solutions Limited, using Jacobsen HKR technology.

Specifications

CAS 352534-75-9
Density 1.018 g/mL at 25°C
Boiling Point 65°C
Refractive Index n20/D 1.535
MDL Number MFCD00282813
Quantity 1 g
Synonym (S)-(-)-Phenyloxirane; (S)-Phenylethylene oxide
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.