Learn More
Fine Science Tools S&T FORCEPS, STRAIGHT
SDP
Catalog No. NC0537033
Encompass
Style: JF-5TC/ Tip Shape: Straight/ Tips: SuperGrip/ Tip Dimensions: 0.3 x 0.25mm/ Alloy / Material: Inox/ Length: 11cm
Catalog No. NC0537033 Supplier Fine Science Tools Supplier No. 0063211
Style: JF-5TC/ Tip Shape: Straight/ Tips: SuperGrip/ Tip Dimensions: 0.3 x 0.25mm/ Alloy / Material: Inox/ Length: 11cm