Learn More
Fine Science Tools S&T VESSEL FORCEPS
SDP
Catalog No. NC9551624
Encompass
Product: S&T Vessel Dilating Forceps- Angled 45 deg./13.5cm/0.2mm Tip Diameter/ Tip Shape: Angled 45 deg./ Tip Diameter: 0.2mm/ Alloy: Stainless Steel/ Length: 13.5cm/ Unit: Each
Catalog No. NC9551624 Supplier Fine Science Tools Supplier No. 0027613
Product: S&T Vessel Dilating Forceps- Angled 45 deg./13.5cm/0.2mm Tip Diameter/ Tip Shape: Angled 45 deg./ Tip Diameter: 0.2mm/ Alloy: Stainless Steel/ Length: 13.5cm/ Unit: Each