Learn More
Sino Biological S-tag PAb 20ul (Sino Biological)
Supplier:  Sino Biological 101290T3820
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the S-tag sequence (KETAAAKFERQHMDS), and purified by antigen affinity chromatography.
Catalog No. 50-237-2688