Learn More
Medchemexpress LLC HY-W074930 100mg Medchemexpress, (S)-Tenofovir CAS:147127-19-3 Purity:>98%
SDP
Catalog No. 501970747
Encompass_Preferred
Medchemexpress, HY-W074930 100mg (S)-Tenofovir CAS:147127-19-3 (S)-Tenofovir ((S)-GS 1278) is the less active S-enantiomer of Tenofovir. Tenofovir is a nucleotide reverse transcriptase inhibitor to treat HIV and chronic Hepatitis B (HBV). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0747 Supplier Medchemexpress LLC Supplier No. HYW074930100MG
Medchemexpress, HY-W074930 100mg (S)-Tenofovir CAS:147127-19-3 (S)-Tenofovir ((S)-GS 1278) is the less active S-enantiomer of Tenofovir. Tenofovir is a nucleotide reverse transcriptase inhibitor to treat HIV and chronic Hepatitis B (HBV). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.