Learn More
Medchemexpress LLC (S)-Terazosin | 109351-33-9 | 99.7% | 100 MG
SDP
Catalog No. 5000406459
Encompass_Preferred
(S)-Terazosin is the active S-enantiomer of Terazosin. It functions as a potent and high-affinity α-adrenoceptor antagonist, demonstrating specific Ki values for α1a, α1b, and α1d-adrenoceptors, as well as high affinity for α2a, α2B, and α2c-adrenoceptors. Its mechanism involves antagonism of α1-adrenoceptor subtypes in isolated tissues and antagonism at rat atrial α2B and rat vas deferens α1A and α2A receptors.
  • Active S-enantiomer of Terazosin
  • Potent α-adrenoceptor antagonist
  • High-affinity for α1a, α1b, and α1d-adrenoceptors
  • High-affinity for α2a, α2B, and α2c-adrenoceptors
  • Antagonism at rat atrial α2B receptor
  • Antagonism at rat vas deferens α1A and α2A receptors
Catalog No. 50-004-06459 Supplier Medchemexpress LLC Supplier No. HYB0371D100MG
(S)-Terazosin is the active S-enantiomer of Terazosin. It functions as a potent and high-affinity α-adrenoceptor antagonist, demonstrating specific Ki values for α1a, α1b, and α1d-adrenoceptors, as well as high affinity for α2a, α2B, and α2c-adrenoceptors. Its mechanism involves antagonism of α1-adrenoceptor subtypes in isolated tissues and antagonism at rat atrial α2B and rat vas deferens α1A and α2A receptors.
  • Active S-enantiomer of Terazosin
  • Potent α-adrenoceptor antagonist
  • High-affinity for α1a, α1b, and α1d-adrenoceptors
  • High-affinity for α2a, α2B, and α2c-adrenoceptors
  • Antagonism at rat atrial α2B receptor
  • Antagonism at rat vas deferens α1A and α2A receptors