Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-Terazosin | 109351-33-9 | 99.7% | 5 MG
SDP

Catalog No. 5000406835
Encompass_Preferred

(S)-Terazosin is the active S-enantiomer of Terazosin, functioning as a potent and high-affinity α-adrenoceptor antagonist. It exhibits specific Ki values for various adrenoceptors, making it suitable for targeted research. This compound is intended for laboratory research use only and not for medical applications.

  • Potent and high-affinity α-adrenoceptor antagonist
  • High affinity for α1a, α1b, α1d, α2B, and α2c-adrenoceptors
  • Shows antagonism of rat atrial α2B receptor
  • Shows antagonism of rat vas deferens α1A and α2A receptor
  • Enantiomeric excess of 99.16%
  • White to off-white solid appearance

Catalog No. 50-004-06835 Supplier Medchemexpress LLC Supplier No. HYB0371D5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-Terazosin is the active S-enantiomer of Terazosin, functioning as a potent and high-affinity α-adrenoceptor antagonist. It exhibits specific Ki values for various adrenoceptors, making it suitable for targeted research. This compound is intended for laboratory research use only and not for medical applications.

  • Potent and high-affinity α-adrenoceptor antagonist
  • High affinity for α1a, α1b, α1d, α2B, and α2c-adrenoceptors
  • Shows antagonism of rat atrial α2B receptor
  • Shows antagonism of rat vas deferens α1A and α2A receptor
  • Enantiomeric excess of 99.16%
  • White to off-white solid appearance

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.