Learn More
Medchemexpress LLC (S)-Terazosin | 109351-33-9 | 99.7% | 50 MG
SDP
Catalog No. 5000405389
Encompass_Preferred
(S)-Terazosin is the active S-enantiomer of Terazosin and a potent, high-affinity α-adrenoceptor antagonist. It exhibits strong binding to α1a, α1b, and α1d-adrenoceptors, as well as significant affinity for α2a, α2B, and α2c-adrenoceptors, making it a powerful antagonist in isolated tissues.
  • Active S-enantiomer of Terazosin.
  • Potent and high-affinity α-adrenoceptor antagonist.
  • Antagonizes α1a, α1b, and α1d-adrenoceptors.
  • Exhibits affinity for α2a, α2B, and α2c-adrenoceptors.
  • Demonstrates antagonism at rat atrial α2B receptor.
  • Antagonizes rat vas deferens α1A and α2A receptors.
Catalog No. 50-004-05389 Supplier Medchemexpress LLC Supplier No. HYB0371D50MG
(S)-Terazosin is the active S-enantiomer of Terazosin and a potent, high-affinity α-adrenoceptor antagonist. It exhibits strong binding to α1a, α1b, and α1d-adrenoceptors, as well as significant affinity for α2a, α2B, and α2c-adrenoceptors, making it a powerful antagonist in isolated tissues.
  • Active S-enantiomer of Terazosin.
  • Potent and high-affinity α-adrenoceptor antagonist.
  • Antagonizes α1a, α1b, and α1d-adrenoceptors.
  • Exhibits affinity for α2a, α2B, and α2c-adrenoceptors.
  • Demonstrates antagonism at rat atrial α2B receptor.
  • Antagonizes rat vas deferens α1A and α2A receptors.