Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-terazosin | 109351-33-9 | 99.9% | 387.43 | C19H25N5O4 | 10 MG
SDP

Catalog No. 5000297458
Encompass_Preferred

(S)-Terazosin is the optically pure S-enantiomer of terazosin and a potent α-adrenoceptor antagonist used in pharmacology and receptor-binding research. It is commonly applied in vitro to characterize receptor subtype affinity and pharmacological activity.

  • Optically pure S-enantiomer for stereoselective studies.
  • Potent α-adrenoceptor antagonist with low-nanomolar Ki values for α1 subtypes.
  • Useful for in vitro pharmacology and receptor-binding assays.
  • Molecular formula C19H25N5O4 and molecular weight 387.43 g/mol.
  • Reported purity 99.9%.
  • Available in small research pack sizes, including 10 mg solid.

Catalog No. 50-002-97458 Supplier Medchemexpress LLC Supplier No. HYB0371D10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-Terazosin is the optically pure S-enantiomer of terazosin and a potent α-adrenoceptor antagonist used in pharmacology and receptor-binding research. It is commonly applied in vitro to characterize receptor subtype affinity and pharmacological activity.

  • Optically pure S-enantiomer for stereoselective studies.
  • Potent α-adrenoceptor antagonist with low-nanomolar Ki values for α1 subtypes.
  • Useful for in vitro pharmacology and receptor-binding assays.
  • Molecular formula C19H25N5O4 and molecular weight 387.43 g/mol.
  • Reported purity 99.9%.
  • Available in small research pack sizes, including 10 mg solid.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.