Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-Tert-Butyl 1-(Chloromethyl)-5-Hydroxy-1H-Benzo[E]Indole-3(2H)-Carboxylate

Catalog No. ALD00034 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ALD00034 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. ALD00034 Supplier Sigma Aldrich Supplier No. ALD0003410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N-Boc-CBI best used as its seco precursor (seco-N-Boc-CBI) is a simple alkylation subunit analog of that found in the naturally occurring antitumor agents the duocarmycins, yatakemycin, and CC-1065, which act by sequence selective DNA alkylation.Design, Synthesis, and Evaluation of Duocarmycin O-Amino Phenol Prodrugs Subject to Tunable Reductive Activation

Specifications

Molecular Formula C7H5Cl2NaO2S
Linear Formula C7H5Cl2NaO2S
Quantity 500 mg
Molecular Weight (g/mol) 247.07 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.