Learn More
Medchemexpress LLC (S)-tert-butyl 1-hydroxy-3-phenylpropan-2-ylcarbamate | 66605-57-0 | 99.9% | C14H21NO3 | 100 G
SDP
Catalog No. 5000433681
Encompass_Preferred
(S)-tert-butyl 1-hydroxy-3-phenylpropan-2-ylcarbamate is a derivative of phenylalanine. This compound is intended for research use only. Amino acids and their derivatives, such as this compound, are utilized as ergogenic supplements, affecting the secretion of anabolic hormones, providing fuel during exercise, and enhancing mental performance during stress. It also helps prevent exercise-induced muscle damage. The product is a white to off-white solid.
  • Derivative of phenylalanine
  • Intended for research use only
  • Acts as an ergogenic supplement
  • Influences anabolic hormone secretion
  • Provides fuel during exercise
  • Enhances mental performance during stress
  • Helps prevent exercise-induced muscle damage
  • Appears as a white to off-white solid
Catalog No. 50-004-33681 Supplier Medchemexpress LLC Supplier No. HY75947100G
(S)-tert-butyl 1-hydroxy-3-phenylpropan-2-ylcarbamate is a derivative of phenylalanine. This compound is intended for research use only. Amino acids and their derivatives, such as this compound, are utilized as ergogenic supplements, affecting the secretion of anabolic hormones, providing fuel during exercise, and enhancing mental performance during stress. It also helps prevent exercise-induced muscle damage. The product is a white to off-white solid.
  • Derivative of phenylalanine
  • Intended for research use only
  • Acts as an ergogenic supplement
  • Influences anabolic hormone secretion
  • Provides fuel during exercise
  • Enhances mental performance during stress
  • Helps prevent exercise-induced muscle damage
  • Appears as a white to off-white solid