Learn More
Medchemexpress LLC (S)-tert-butyl 3-(2-hydroxyethyl)piperazine-1-carboxylate | 1273577-11-9 | MFCD18633590 | 98.1% | 230.31 | C11H22N2O3 | 5 G

Supplier: Medchemexpress LLC HYW0693345G
(S)-tert-butyl 3-(2-hydroxyethyl)piperazine-1-carboxylate is an S-enantiomer carbamate derivative of piperazine used as a drug intermediate in synthetic chemistry and research applications. It is characterized by CAS 1273577-11-9, molecular formula C11H22N2O3, molecular weight 230.31 g/mol, and a reported purity of 98.11% (GC).
- used as a drug intermediate in medicinal chemistry
- high purity: 98.11% (GC)
- molecular formula C11H22N2O3 and molecular weight 230.31 g/mol
- solid form; storage: 4°C and protected from light
- in solvent: store at -80°C up to 6 months, or -20°C up to 1 month (protect from light)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.