Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-tert-butyl 3-(2-hydroxyethyl)piperazine-1-carboxylate | 1273577-11-9 | MFCD18633590 | 98.1% | 230.31 | C11H22N2O3 | 50 G
SDP

Catalog No. 5000210248
Encompass_Preferred

(S)-tert-butyl 3-(2-hydroxyethyl)piperazine-1-carboxylate is a chiral carbamate intermediate used in medicinal chemistry for the synthesis of various active compounds. It is supplied for research use and characterized by analytical data including GC purity and molecular weight.

  • Chiral carbamate intermediate for medicinal chemistry
  • CAS number 1273577-11-9
  • Molecular formula C11H22N2O3
  • Molecular weight 230.31
  • Purity 98.1% (GC)
  • Appearance: liquid; recommended storage 4°C, protect from light
  • Supplied in laboratory-scale quantities (e.g., 50 G)

Catalog No. 50-002-10248 Supplier Medchemexpress LLC Supplier No. HYW06933450G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-tert-butyl 3-(2-hydroxyethyl)piperazine-1-carboxylate is a chiral carbamate intermediate used in medicinal chemistry for the synthesis of various active compounds. It is supplied for research use and characterized by analytical data including GC purity and molecular weight.

  • Chiral carbamate intermediate for medicinal chemistry
  • CAS number 1273577-11-9
  • Molecular formula C11H22N2O3
  • Molecular weight 230.31
  • Purity 98.1% (GC)
  • Appearance: liquid; recommended storage 4°C, protect from light
  • Supplied in laboratory-scale quantities (e.g., 50 G)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.