Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-tert-butyl 3-(2-hydroxyethyl)piperazine-1-carboxylate | 1273577-11-9 | MFCD18633590 | 98.1% | 230.31 g/mol | C11H22N2O3 | 100 G
SDP

Catalog No. 5000210244
Encompass_Preferred

(S)-tert-butyl 3-(2-hydroxyethyl)piperazine-1-carboxylate is a chiral carbamate-protected piperazine derivative used as a synthetic intermediate in medicinal chemistry and organic synthesis. It is supplied for research use only.

  • Chiral intermediate for drug synthesis.
  • Cas number 1273577-11-9.
  • Purity 98.1% (GC).
  • Molecular weight 230.31 g/mol, formula C11H22N2O3.
  • Typically supplied in 100 g packages.
  • Intended for research use only; not for human consumption.

Catalog No. 50-002-10244 Supplier Medchemexpress LLC Supplier No. HYW069334100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-tert-butyl 3-(2-hydroxyethyl)piperazine-1-carboxylate is a chiral carbamate-protected piperazine derivative used as a synthetic intermediate in medicinal chemistry and organic synthesis. It is supplied for research use only.

  • Chiral intermediate for drug synthesis.
  • Cas number 1273577-11-9.
  • Purity 98.1% (GC).
  • Molecular weight 230.31 g/mol, formula C11H22N2O3.
  • Typically supplied in 100 g packages.
  • Intended for research use only; not for human consumption.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.