Learn More
Medchemexpress LLC S-tert-butyl-L-cysteine hydrochloride | 2481-09-6 | MFCD00037426 | 98.0% | 213.73 g/mol | C7H16ClNO2S | 100 G
SDP
Catalog No. 5000304190
Encompass_Preferred
S-tert-butyl-L-cysteine hydrochloride is a tert-butyl-protected cysteine hydrochloride salt used as an amino acid building block in peptide and small-molecule synthesis. The protected thiol enables selective transformations on the amino acid backbone and facilitates incorporation into synthetic sequences. The product is supplied as a solid with defined purity and storage recommendations for long-term stability.
  • Protected cysteine derivative suitable for peptide and organic synthesis.
  • Hydrochloride salt form for improved handling and stability.
  • High purity (98.0%) appropriate for research applications.
  • Molecular weight 213.73 g/mol; formula C7H16ClNO2S.
  • Available in multiple package sizes, including 100 g.
  • Stable solid with recommended cold-storage conditions for long-term storage.
Catalog No. 50-003-04190 Supplier Medchemexpress LLC Supplier No. HYW014238100G
S-tert-butyl-L-cysteine hydrochloride is a tert-butyl-protected cysteine hydrochloride salt used as an amino acid building block in peptide and small-molecule synthesis. The protected thiol enables selective transformations on the amino acid backbone and facilitates incorporation into synthetic sequences. The product is supplied as a solid with defined purity and storage recommendations for long-term stability.
  • Protected cysteine derivative suitable for peptide and organic synthesis.
  • Hydrochloride salt form for improved handling and stability.
  • High purity (98.0%) appropriate for research applications.
  • Molecular weight 213.73 g/mol; formula C7H16ClNO2S.
  • Available in multiple package sizes, including 100 g.
  • Stable solid with recommended cold-storage conditions for long-term storage.