Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-tert-butyl ... piperazine-1-carboxylate formate | 00-00-0 | 98.9% | 835.0 g/mol | C43H62N8O9 | 5 MG
SDP

Catalog No. 5000262924
Encompass_Preferred

K-Ras ligand-Linker Conjugate 6 formate is a target-protein ligand-linker conjugate provided as the formate salt for use as a building block in PROTAC synthesis and KRAS-targeted chemical biology research. The material is supplied at research scale and is characterized by a certificate of analysis that documents identity and purity.

  • Intended for PROTAC synthesis and KRAS-targeted research.
  • Formate salt isolated and characterized by batch COA.
  • High purity suitable for synthetic use (98.9%).
  • Molecular weight 835.0 g/mol and formula C43H62N8O9 documented.
  • Available in small research-scale pack sizes (5 mg).

Catalog No. 50-002-62924 Supplier Medchemexpress LLC Supplier No. HY130991A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

K-Ras ligand-Linker Conjugate 6 formate is a target-protein ligand-linker conjugate provided as the formate salt for use as a building block in PROTAC synthesis and KRAS-targeted chemical biology research. The material is supplied at research scale and is characterized by a certificate of analysis that documents identity and purity.

  • Intended for PROTAC synthesis and KRAS-targeted research.
  • Formate salt isolated and characterized by batch COA.
  • High purity suitable for synthetic use (98.9%).
  • Molecular weight 835.0 g/mol and formula C43H62N8O9 documented.
  • Available in small research-scale pack sizes (5 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.