Learn More
Medchemexpress LLC (S)-(-)-tert-Butylsulfinamide | 343338-28-3 | 99.9% | 121.20 | 25 G
SDP
Catalog No. 5000402379
Encompass_Preferred
(S)-(-)-tert-Butylsulfinamide is a compound featuring a chiral sulfur atom. It is utilized in the synthesis of other compounds and serves as a chiral auxiliary in the asymmetric synthesis of organic compounds.
  • Contains a chiral sulfur atom
  • Used in the synthesis of other compounds
  • Acts as a chiral auxiliary in asymmetric synthesis of organic compounds
Catalog No. 50-004-02379 Supplier Medchemexpress LLC Supplier No. 25GHY40229
(S)-(-)-tert-Butylsulfinamide is a compound featuring a chiral sulfur atom. It is utilized in the synthesis of other compounds and serves as a chiral auxiliary in the asymmetric synthesis of organic compounds.
  • Contains a chiral sulfur atom
  • Used in the synthesis of other compounds
  • Acts as a chiral auxiliary in asymmetric synthesis of organic compounds