Learn More
Medchemexpress LLC (S)-thalidomide-4-oh | 2357105-36-1 | 98.0% | 274.23 g/mol | C13H10N2O5 | 50 MG
SDP
Catalog No. 5000284948
Encompass_Preferred
(S)-Thalidomide-4-OH is the S-enantiomer of thalidomide-4-OH, a thalidomide-derived cereblon ligand used in chemical biology and medicinal chemistry. Supplied as a solid research reagent, it is commonly used to recruit CRBN in PROTAC design and as a building block for medicinal chemistry.
  • S-enantiomer of thalidomide-4-OH for stereospecific studies.
  • Cereblon (CRBN) ligand for E3 ligase recruitment.
  • Suitable building block for PROTAC synthesis and linker chemistry.
  • Off-white to light yellow solid; typical pack size 50 mg.
  • Reported purity 98.0% and molecular weight 274.23 g/mol.
Catalog No. 50-002-84948 Supplier Medchemexpress LLC Supplier No. HYW67443650MG
(S)-Thalidomide-4-OH is the S-enantiomer of thalidomide-4-OH, a thalidomide-derived cereblon ligand used in chemical biology and medicinal chemistry. Supplied as a solid research reagent, it is commonly used to recruit CRBN in PROTAC design and as a building block for medicinal chemistry.
  • S-enantiomer of thalidomide-4-OH for stereospecific studies.
  • Cereblon (CRBN) ligand for E3 ligase recruitment.
  • Suitable building block for PROTAC synthesis and linker chemistry.
  • Off-white to light yellow solid; typical pack size 50 mg.
  • Reported purity 98.0% and molecular weight 274.23 g/mol.