Learn More
Medchemexpress LLC S-thalidomide-4-OH | 2357105-36-1 | MFCD03699892 | 98.0% | 274.23 | C13H10N2O5 | 25 MG
SDP
Catalog No. 5000284164
Encompass_Preferred
(S)-Thalidomide-4-OH is the S-enantiomer of thalidomide-4-OH, a thalidomide-based cereblon (CRBN) ligand used in chemical biology and proteolysis-targeting chimera (PROTAC) design. Supplied as a high-purity powder, it is intended for research use in CRBN recruitment studies and degrader synthesis.
  • High purity (98.0%) for research applications.
  • S-enantiomer for stereospecific biochemical studies.
  • Useful cereblon ligand for PROTAC conjugation.
  • Available in small milligram quantities for screening and synthesis.
  • Stable when stored under recommended conditions.
Catalog No. 50-002-84164 Supplier Medchemexpress LLC Supplier No. HYW67443625MG
(S)-Thalidomide-4-OH is the S-enantiomer of thalidomide-4-OH, a thalidomide-based cereblon (CRBN) ligand used in chemical biology and proteolysis-targeting chimera (PROTAC) design. Supplied as a high-purity powder, it is intended for research use in CRBN recruitment studies and degrader synthesis.
  • High purity (98.0%) for research applications.
  • S-enantiomer for stereospecific biochemical studies.
  • Useful cereblon ligand for PROTAC conjugation.
  • Available in small milligram quantities for screening and synthesis.
  • Stable when stored under recommended conditions.