Learn More
TARGETMOL CHEMICALS INC S-THALIDOMIDE 5MG
SDP
Catalog No. 502577419
Encompass_Preferred
Also available in 10mg 25mg 50mg 100mg 500mg and bulk. Please contact Fisher for quotes. (S)-Thalidomide ((S)-(-)-Thalidomide) is the S-isomer of Thalidomide with immunomodulatory anti-inflammatory anticancer anti-angiogenic and pro-apoptotic activities and is used in the study of leprosy erythema nodosum and myeloma. purity: 99%
Catalog No. 50-257-7419 Supplier TARGETMOL CHEMICALS INC Supplier No. T12644L5MG
Also available in 10mg 25mg 50mg 100mg 500mg and bulk. Please contact Fisher for quotes. (S)-Thalidomide ((S)-(-)-Thalidomide) is the S-isomer of Thalidomide with immunomodulatory anti-inflammatory anticancer anti-angiogenic and pro-apoptotic activities and is used in the study of leprosy erythema nodosum and myeloma. purity: 99%