Learn More
Medchemexpress LLC (S)-Thalidomide | 841-67-8 | 99.5% | 258.23 | 100 MG
SDP
Catalog No. 5000360720
Encompass_Preferred
(S)-Thalidomide is the S-enantiomer of Thalidomide. It possesses immunomodulatory, anti-inflammatory, antiangiogenic, and pro-apoptotic effects. (S)-Thalidomide induces teratogenic effects by binding to cereblon (CRBN).
  • Immunomodulatory effects
  • Anti-inflammatory effects
  • Antiangiogenic effects
  • Pro-apoptotic effects
  • Induces teratogenic effects by binding to cereblon (CRBN)
  • For research use only
Catalog No. 50-003-60720 Supplier Medchemexpress LLC Supplier No. HY14658A100MG
(S)-Thalidomide is the S-enantiomer of Thalidomide. It possesses immunomodulatory, anti-inflammatory, antiangiogenic, and pro-apoptotic effects. (S)-Thalidomide induces teratogenic effects by binding to cereblon (CRBN).
  • Immunomodulatory effects
  • Anti-inflammatory effects
  • Antiangiogenic effects
  • Pro-apoptotic effects
  • Induces teratogenic effects by binding to cereblon (CRBN)
  • For research use only