Learn More
Medchemexpress LLC (S)-thalidomide | 841-67-8 | MFCD00210219 | 99.5% | C13H10N2O4 | 10MG
SDP
Catalog No. 5000259413
Encompass_Preferred
(S)-Thalidomide is the S-enantiomer of thalidomide, a small-molecule ligand of the E3 ubiquitin ligase substrate receptor cereblon (CRBN). It exhibits immunomodulatory, anti-inflammatory, antiangiogenic, and pro-apoptotic activities and is used as a research reagent to study CRBN-mediated degradation pathways and molecular glue mechanisms. For research use only.
  • High chemical purity suitable for analytical and biological studies.
  • Known to bind cereblon and induce substrate recruitment for degradation.
  • Useful in studies of immunomodulation, angiogenesis, and apoptosis.
  • Available in small milligram pack sizes for screening and synthesis.
  • Provided with manufacturer documentation including SDS and COA.
Catalog No. 50-002-59413 Supplier Medchemexpress LLC Supplier No. HY14658A10MG
(S)-Thalidomide is the S-enantiomer of thalidomide, a small-molecule ligand of the E3 ubiquitin ligase substrate receptor cereblon (CRBN). It exhibits immunomodulatory, anti-inflammatory, antiangiogenic, and pro-apoptotic activities and is used as a research reagent to study CRBN-mediated degradation pathways and molecular glue mechanisms. For research use only.
  • High chemical purity suitable for analytical and biological studies.
  • Known to bind cereblon and induce substrate recruitment for degradation.
  • Useful in studies of immunomodulation, angiogenesis, and apoptosis.
  • Available in small milligram pack sizes for screening and synthesis.
  • Provided with manufacturer documentation including SDS and COA.