Learn More
Medchemexpress LLC (S)-Thalidomide | 841-67-8 | MFCD00210219 | 99.52% | 258.23 | 50 MG
SDP
Catalog No. 5000360721
Encompass_Preferred
(S)-Thalidomide is the S-enantiomer of Thalidomide. It exhibits immunomodulatory, anti-inflammatory, antiangiogenic, and pro-apoptotic effects. (S)-Thalidomide induces teratogenic effects by binding to cereblon (CRBN).
  • It is the S-enantiomer of thalidomide.
  • It exhibits immunomodulatory effects.
  • It has anti-inflammatory properties.
  • It shows antiangiogenic effects.
  • It possesses pro-apoptotic effects.
  • It binds to cereblon (CRBN).
Catalog No. 50-003-60721 Supplier Medchemexpress LLC Supplier No. HY14658A50MG
(S)-Thalidomide is the S-enantiomer of Thalidomide. It exhibits immunomodulatory, anti-inflammatory, antiangiogenic, and pro-apoptotic effects. (S)-Thalidomide induces teratogenic effects by binding to cereblon (CRBN).
  • It is the S-enantiomer of thalidomide.
  • It exhibits immunomodulatory effects.
  • It has anti-inflammatory properties.
  • It shows antiangiogenic effects.
  • It possesses pro-apoptotic effects.
  • It binds to cereblon (CRBN).