Learn More
Medchemexpress LLC (S)-thioctic acid | 1077-27-6 | MFCD01863481 | >=97.0% | 206.33 g/mol | C8H14O2S2 | 1 ML
SDP
Catalog No. 5000302747
Encompass_Preferred
(S)-Thioctic acid (S-lipoic acid) is the S-enantiomer of lipoic acid, a thiol-containing antioxidant and enzymatic cofactor used in biochemical research. It participates in redox chemistry and mitochondrial enzyme function. The product is supplied as a ready-to-use 10 mM solution in DMSO and as solids for preparative or analytical work, suitable for enzymology and antioxidant studies.
  • Chiral antioxidant and mitochondrial cofactor
  • Available as 10 mM solution in DMSO for immediate use
  • Also available as solid for bulk or formulation
  • Useful for enzymology, redox, and antioxidant assays
  • High purity (≥97%) suitable for HPLC applications
  • Stable when stored under recommended conditions
Catalog No. 50-003-02747 Supplier Medchemexpress LLC Supplier No. HYW01459010MM/1ML
(S)-Thioctic acid (S-lipoic acid) is the S-enantiomer of lipoic acid, a thiol-containing antioxidant and enzymatic cofactor used in biochemical research. It participates in redox chemistry and mitochondrial enzyme function. The product is supplied as a ready-to-use 10 mM solution in DMSO and as solids for preparative or analytical work, suitable for enzymology and antioxidant studies.
  • Chiral antioxidant and mitochondrial cofactor
  • Available as 10 mM solution in DMSO for immediate use
  • Also available as solid for bulk or formulation
  • Useful for enzymology, redox, and antioxidant assays
  • High purity (≥97%) suitable for HPLC applications
  • Stable when stored under recommended conditions