Learn More
Medchemexpress LLC (S)-thioctic acid | 1077-27-6 | MFCD01863481 | 99.8% | 206.33 | C8H14O2S2 | 5 G
SDP
Catalog No. 5000303948
Encompass_Preferred
(S)-Thioctic acid is the S-enantiomer of lipoic (thioctic) acid supplied as a research-grade biochemical reagent. It acts as an organosulfur antioxidant and a cofactor analogue, suitable for biochemical assays, stereospecific studies, and analytical characterization.
  • High chemical purity (99.79%).
  • Chiral S-enantiomer for stereospecific studies.
  • Solid form, light yellow to yellow appearance.
  • Suitable for biochemical assays and antioxidant research.
  • Powder storage: -20°C (3 years) or 4°C (2 years).
  • Includes COA, SDS, and spectral data for quality verification.
Catalog No. 50-003-03948 Supplier Medchemexpress LLC Supplier No. HYW0145905G
(S)-Thioctic acid is the S-enantiomer of lipoic (thioctic) acid supplied as a research-grade biochemical reagent. It acts as an organosulfur antioxidant and a cofactor analogue, suitable for biochemical assays, stereospecific studies, and analytical characterization.
  • High chemical purity (99.79%).
  • Chiral S-enantiomer for stereospecific studies.
  • Solid form, light yellow to yellow appearance.
  • Suitable for biochemical assays and antioxidant research.
  • Powder storage: -20°C (3 years) or 4°C (2 years).
  • Includes COA, SDS, and spectral data for quality verification.