Learn More
Medchemexpress LLC (S)-Timolol (Maleate) | 26921-17-5 | 99.8% | 432.49 | 10 MM * 1 ML
SDP
Catalog No. 5000386008
Encompass_Preferred
(S)-Timolol Maleate is a non-cardioselective hydrophilic β-adrenoceptor blocker. It is commonly used as a standard medication for intraocular pressure (glaucoma) by preventing the production of aqueous humor. It can also be used for hypertension, angina pectoris, and myocardial infarction.
  • Non-cardioselective hydrophilic β-adrenoceptor blocker
  • Prevents the production of aqueous humor
  • Used for intraocular pressure
  • Effective in hypertension, angina pectoris, and myocardial infarction
Catalog No. 50-003-86008 Supplier Medchemexpress LLC Supplier No. HY1738010MM/1ML
(S)-Timolol Maleate is a non-cardioselective hydrophilic β-adrenoceptor blocker. It is commonly used as a standard medication for intraocular pressure (glaucoma) by preventing the production of aqueous humor. It can also be used for hypertension, angina pectoris, and myocardial infarction.
  • Non-cardioselective hydrophilic β-adrenoceptor blocker
  • Prevents the production of aqueous humor
  • Used for intraocular pressure
  • Effective in hypertension, angina pectoris, and myocardial infarction