Learn More
Medchemexpress LLC (S)-Timolol maleate | 26921-17-5 | 99.8% | 432.49 | 200 MG
SDP
Catalog No. 5000384776
Encompass_Preferred
(S)-Timolol Maleate (L-714,465 Maleate) is a non-cardioselective hydrophilic β-adrenoceptor blocker. It is widely used as standard medication for intraocular pressure (glaucoma) by preventing the production of aqueous humor. It can also be used for hypertension, angina pectoris, and myocardial infarction.
  • Non-cardioselective β-adadrenergic blocker
  • Used for intraocular pressure (glaucoma)
  • Prevents production of aqueous humor
  • Suitable for hypertension
  • Suitable for angina pectoris
  • Suitable for myocardial infarction
Catalog No. 50-003-84776 Supplier Medchemexpress LLC Supplier No. HY17380200MG
(S)-Timolol Maleate (L-714,465 Maleate) is a non-cardioselective hydrophilic β-adrenoceptor blocker. It is widely used as standard medication for intraocular pressure (glaucoma) by preventing the production of aqueous humor. It can also be used for hypertension, angina pectoris, and myocardial infarction.
  • Non-cardioselective β-adadrenergic blocker
  • Used for intraocular pressure (glaucoma)
  • Prevents production of aqueous humor
  • Suitable for hypertension
  • Suitable for angina pectoris
  • Suitable for myocardial infarction