Learn More
Medchemexpress LLC (S)-Timolol (Maleate) | 26921-17-5 | 99.8% | 432.49 | 25 MG
SDP
Catalog No. 5000384128
Encompass_Preferred
(S)-Timolol maleate is a non-cardioselective hydrophilic β-adrenoceptor blocker, widely used as a standard medication for intraocular pressure (glaucoma) by preventing the production of aqueous humor. It also finds application in hypertension, angina pectoris, and myocardial infarction. This product is suitable for research and analytical applications.
  • Non-cardioselective hydrophilic β-adrenoceptor blocker
  • Prevents production of aqueous humor
  • Used for intraocular pressure (glaucoma)
  • Can be used for hypertension
  • Can be used for angina pectoris
  • Can be used for myocardial infarction
Catalog No. 50-003-84128 Supplier Medchemexpress LLC Supplier No. 25MGHY17380R
(S)-Timolol maleate is a non-cardioselective hydrophilic β-adrenoceptor blocker, widely used as a standard medication for intraocular pressure (glaucoma) by preventing the production of aqueous humor. It also finds application in hypertension, angina pectoris, and myocardial infarction. This product is suitable for research and analytical applications.
  • Non-cardioselective hydrophilic β-adrenoceptor blocker
  • Prevents production of aqueous humor
  • Used for intraocular pressure (glaucoma)
  • Can be used for hypertension
  • Can be used for angina pectoris
  • Can be used for myocardial infarction