Learn More
Medchemexpress LLC (S)-Timolol maleate | 26921-17-5 | 99.8% | 432.49 | 50MG
SDP
Catalog No. 5000384575
Encompass_Preferred
(S)-Timolol maleate is a non-cardioselective hydrophilic β-adrenoceptor blocker, commonly used as a standard medication for intraocular pressure (glaucoma) by preventing the production of aqueous humor. It can also be used for hypertension, angina pectoris, and myocardial infarction. As a chiral compound with one asymmetric carbon, the (S)-enantiomer is a non-cardioselective β-adrenergic blocker, with the (R)-isomer having lower biological activity.
  • Non-cardioselective hydrophilic β-adrenoceptor blocker
  • Used for intraocular pressure (glaucoma)
  • Used for hypertension, angina pectoris, and myocardial infarction
  • Single (S)-enantiomer
Catalog No. 50-003-84575 Supplier Medchemexpress LLC Supplier No. HY17380R50MG
(S)-Timolol maleate is a non-cardioselective hydrophilic β-adrenoceptor blocker, commonly used as a standard medication for intraocular pressure (glaucoma) by preventing the production of aqueous humor. It can also be used for hypertension, angina pectoris, and myocardial infarction. As a chiral compound with one asymmetric carbon, the (S)-enantiomer is a non-cardioselective β-adrenergic blocker, with the (R)-isomer having lower biological activity.
  • Non-cardioselective hydrophilic β-adrenoceptor blocker
  • Used for intraocular pressure (glaucoma)
  • Used for hypertension, angina pectoris, and myocardial infarction
  • Single (S)-enantiomer