Learn More
Medchemexpress LLC (S)-Timolol maleate | 26921-17-5 | 99.9% | 432.49 | 100 MG
SDP
Catalog No. 5000364867
Encompass_Preferred
(S)-Timolol Maleate (L-714,465 Maleate) is a non-cardioselective hydrophilic β-adrenoceptor blocker. It is widely used as a standard medication for intraocular pressure (glaucoma) by preventing the production of aqueous humor. This compound can also be used for hypertension, angina pectoris, and myocardial infarction.
  • Non-cardioselective hydrophilic β-adrenoceptor blocker
  • Used as standard medication for intraocular pressure (glaucoma)
  • Prevents the production of aqueous humor
  • Can be used for hypertension, angina pectoris, and myocardial infarction
Catalog No. 50-003-64867 Supplier Medchemexpress LLC Supplier No. 100MGHY17380
(S)-Timolol Maleate (L-714,465 Maleate) is a non-cardioselective hydrophilic β-adrenoceptor blocker. It is widely used as a standard medication for intraocular pressure (glaucoma) by preventing the production of aqueous humor. This compound can also be used for hypertension, angina pectoris, and myocardial infarction.
  • Non-cardioselective hydrophilic β-adrenoceptor blocker
  • Used as standard medication for intraocular pressure (glaucoma)
  • Prevents the production of aqueous humor
  • Can be used for hypertension, angina pectoris, and myocardial infarction