Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-timolol maleate | 26921-17-5 | MFCD00058356 | 100.0% | 432.49 | C17H28N4O7S | 10 MG
SDP

Catalog No. 5000299093
Encompass_Preferred

(S)-Timolol maleate is the S-enantiomer of timolol supplied as the maleate salt and provided as an analytical/research standard for pharmacological testing, method development, and quality control. It is a non-cardioselective, hydrophilic β-adrenoceptor blocker commonly used in research related to intraocular pressure and cardiovascular effects.

  • Analytical standard for method development and quality control.
  • S-enantiomer maleate salt with defined stereochemistry.
  • High purity suitable for HPLC and reference standard use.
  • Molecular weight 432.49 and formula C17H28N4O7S.
  • Store sealed at 4°C, protected from light and moisture.

Catalog No. 50-002-99093 Supplier Medchemexpress LLC Supplier No. HY17380R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-Timolol maleate is the S-enantiomer of timolol supplied as the maleate salt and provided as an analytical/research standard for pharmacological testing, method development, and quality control. It is a non-cardioselective, hydrophilic β-adrenoceptor blocker commonly used in research related to intraocular pressure and cardiovascular effects.

  • Analytical standard for method development and quality control.
  • S-enantiomer maleate salt with defined stereochemistry.
  • High purity suitable for HPLC and reference standard use.
  • Molecular weight 432.49 and formula C17H28N4O7S.
  • Store sealed at 4°C, protected from light and moisture.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.