Learn More
Medchemexpress LLC (S)-Timolol maleate (Standard) | 26921-17-5 | 99.99% | 432.49 | 100 MG
SDP
Catalog No. 5000386009
Encompass_Preferred
(S)-Timolol maleate (Standard) is the analytical standard of (S)-Timolol (maleate), intended for research and analytical applications. It is a non-cardioselective hydrophilic β-adrenoceptor blocker widely used as standard medication for intraocular pressure (glaucoma) by preventing the production of aqueous humor. It can also be used for hypertension, angina pectoris, and myocardial infarction.
  • Reference standard supplied assay
  • Commonly used in qualitative, quantitative and methodological research experiments
  • Suitable for HPLC, GC and MS applications
Catalog No. 50-003-86009 Supplier Medchemexpress LLC Supplier No. HY17380R100MG
(S)-Timolol maleate (Standard) is the analytical standard of (S)-Timolol (maleate), intended for research and analytical applications. It is a non-cardioselective hydrophilic β-adrenoceptor blocker widely used as standard medication for intraocular pressure (glaucoma) by preventing the production of aqueous humor. It can also be used for hypertension, angina pectoris, and myocardial infarction.
  • Reference standard supplied assay
  • Commonly used in qualitative, quantitative and methodological research experiments
  • Suitable for HPLC, GC and MS applications