Learn More
Hach Company Simplified TKN (s-TKN) TNTplus Vial Test (0-16 mg/L N), 25 Tests
Catalog No. 501998164 Shop All Hach Company Products
Encompass
Total Kjeldahl Nitrogen TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime.Simplified TKN (s-TKN™) is method accepted for compliance reporting in many states. Please check with local regulators for acceptance in your state.Hach Method 10242 is compliant for EPA wastewater reporting and gives three results for each test - Total Nitrogen, combined nitrate and nitrite, as well as TKN. Unique vial reagent system digests the sample at only 100 °C, increasing safety while eliminating ammonia contamination and the need to clean glassware. TNT880 has a range up to 16 mg/L TKN.
Catalog No. 50-199-8164 Supplier Hach Company Supplier No. TNT880
Total Kjeldahl Nitrogen TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime.Simplified TKN (s-TKN™) is method accepted for compliance reporting in many states. Please check with local regulators for acceptance in your state.Hach Method 10242 is compliant for EPA wastewater reporting and gives three results for each test - Total Nitrogen, combined nitrate and nitrite, as well as TKN. Unique vial reagent system digests the sample at only 100 °C, increasing safety while eliminating ammonia contamination and the need to clean glassware. TNT880 has a range up to 16 mg/L TKN.